Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium tetrakis(perfluorophenyl)borate 98%

Supplier: AMBEED, INC
Warning

A310435-100MG A310435-1G A310435-250MG
77167-102EA 15 USD InStock
77167-102 77167-104 77167-106
Sodium tetrakis(perfluorophenyl)borate 98%
Sodium tetrakis(perfluorophenyl)borate
Precaución: for Research Use Only


Formula: C₂₄BF₂₀Na
MW: 702.02 g/mol
MDL Number: MFCD12545881
CAS Number: 149213-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty