Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3,3'-Bis(3,5-di-tert-butylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%

Supplier: AMBEED, INC
Warning

A310796-100MG A310796-250MG A310796-50MG
77194-622EA 147 USD InStock
77194-622 77194-624 77194-626
(S)-3,3'-Bis(3,5-di-tert-butylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%
(S)-3,3'-Bis(3,5-di-tert-butylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Precaución: for Research Use Only


Formula: C₄₈H₅₃O₄P
MDL Number: MFCD28139326
CAS Number: 1442645-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty