Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-3-nitrophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A311294-100MG A311294-1G A311294-250MG A311294-5G
77194-644EA 12 USD InStock
77194-644 77194-646 77194-648 77194-650
4-Methoxy-3-nitrophenylboronic acid 98%
4-Methoxy-3-nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BNO₅
MDL Number: MFCD03092922
CAS Number: 827614-67-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty