Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Fluorophenyl)cyclopropanamine hydrochloride 96%

Supplier: AMBEED, INC
Warning

A311838-100MG A311838-1G A311838-250MG
77167-138EA 23 USD InStock
77167-138 77167-140 77167-142
1-(4-Fluorophenyl)cyclopropanamine hydrochloride 96%
1-(4-Fluorophenyl)cyclopropanamine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₁ClFN
MW: 187.64 g/mol
MDL Number: MFCD07995728
CAS Number: 1134524-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty