Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoropiperidine-4-carboxylic acid, N-BOC protected 98%

Supplier: AMBEED, INC
Warning

A313190-100MG A313190-1G A313190-250MG
77194-690EA 7 USD InStock
77194-690 77194-692 77194-694
4-Fluoropiperidine-4-carboxylic acid, N-BOC protected 98%
4-Fluoropiperidine-4-carboxylic acid, N-BOC protected
Precaución: for Research Use Only


Formula: C₁₁H₁₈FNO₄
MDL Number: MFCD08062530
CAS Number: 614731-04-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty