Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-dioxopyrrolidin-1-yldodecanoate 97%

Supplier: AMBEED, INC
Warning

A316427-10G A316427-1G A316427-250MG A316427-5G
77194-764EA 161 USD InStock
77194-764 77194-766 77194-768 77194-770
2,5-dioxopyrrolidin-1-yldodecanoate 97%
2,5-dioxopyrrolidin-1-yldodecanoate
Precaución: for Research Use Only


Formula: C₁₆H₂₇NO₄
MDL Number: MFCD00050401
CAS Number: 14565-47-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty