Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-tert-Butyl 3-methyl 1H-indole-1,3-dicarboxylate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A319771-1G A319771-250MG A319771-5G
77167-266EA 24 USD InStock
77167-266 77167-268 77683-950
1-tert-Butyl 3-methyl 1H-indole-1,3-dicarboxylate 97%, Ambeed.Inc
1-tert-Butyl 3-methyl 1H-indole-1,3-dicarboxylate
Precaución: For research use only.


Formula: C₁₅H₁₇NO₄
MDL Number: MFCD06658476
CAS Number: 338760-26-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty