Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-tert-Butyl-1H-imidazole-4-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A321529-100MG A321529-1G A321529-250MG A321529-5G
77167-284EA 6 USD InStock
77167-284 77167-286 77167-288 77167-290
5-tert-Butyl-1H-imidazole-4-carbaldehyde 95%
5-tert-Butyl-1H-imidazole-4-carbaldehyde
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O
MDL Number: MFCD14583325
CAS Number: 714273-83-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty