Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-4-bromo-1H-imidazole 98%

Supplier: AMBEED, INC
Warning

A321928-100MG A321928-1G A321928-250MG
77194-842EA 100 USD InStock
77194-842 77194-844 77194-846
1-Benzyl-4-bromo-1H-imidazole 98%
1-Benzyl-4-bromo-1H-imidazole
Precaución: for Research Use Only


Formula: C₁₀H₉BrN₂
MDL Number: MFCD09033094
CAS Number: 106848-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty