Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(2-Methylpyridin-4-yl)phenyl)-N-4-(pyridin-3-yl)phenyl)acetamide 98%

Supplier: AMBEED, INC
Warning

A321955-10MG A321955-1MG A321955-5MG
77167-304EA 73 USD InStock
77167-304 77167-306 77167-308
2-(4-(2-Methylpyridin-4-yl)phenyl)-N-4-(pyridin-3-yl)phenyl)acetamide 98%
2-(4-(2-Methylpyridin-4-yl)phenyl)-N-4-(pyridin-3-yl)phenyl)acetamide
Precaución: for Research Use Only


Formula: C₂₅H₂₁N₃O
MDL Number: MFCD22201167
CAS Number: 1243243-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty