Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,3-dimethoxybenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A322477-1G A322477-250MG A322477-5G
77167-330EA 25 USD InStock
77167-330 77167-332 77167-334
5-Chloro-2,3-dimethoxybenzaldehyde 95%
5-Chloro-2,3-dimethoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₉ClO₃
MDL Number: MFCD01743107
CAS Number: 86232-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty