Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrobenzylamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A324728-10G A324728-1G A324728-25G A324728-5G
77194-900EA 95 USD InStock
77194-900 77194-902 77194-904 77194-906
2-Nitrobenzylamine hydrochloride 97%
2-Nitrobenzylamine hydrochloride


Formula: C₇H₈N₂O₂·HCl
MDL Number: MFCD00136280
CAS Number: 24835-08-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty