Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Benzenedimethanethiol 98%

Supplier: AMBEED, INC
Danger

A327563-1G A327563-250MG A327563-25G A327563-5G
77194-930EA 35 USD InStock
77194-930 77194-932 77194-934 77194-936
1,3-Benzenedimethanethiol 98%
1,3-Benzenedimethanethiol
Precaución: for Research Use Only


Formula: C₈H₁₀S₂
MW: 170.29 g/mol
MDL Number: MFCD00012311
CAS Number: 41563-69-3
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty