Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Aminomethyl)phenol acetic acid salt 95%

Supplier: AMBEED, INC
Warning

A328350-100MG A328350-1G A328350-250MG A328350-5G
77194-948EA 5 USD InStock
77194-948 77194-950 77194-952 77194-954
2-(Aminomethyl)phenol acetic acid salt 95%
2-(Aminomethyl)phenol acetic acid salt
Precaución: for Research Use Only


Formula: C₉H₁₃NO₃
MDL Number: MFCD29059387
CAS Number: 1206675-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty