Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-7-iodofluorene 98%

Supplier: AMBEED, INC
Warning

A329512-100G A329512-10G A329512-25G A329512-500G A329512-5G
77167-396EA 63 USD InStock
77167-396 77167-398 77167-400 77167-402 77167-404
2-Bromo-7-iodofluorene 98%
2-Bromo-7-iodofluorene
Precaución: for Research Use Only


Formula: C₁₃H₈BrI
MDL Number: MFCD12024276
CAS Number: 123348-27-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty