Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Mesityl-2,7-dimethyl-10-phenylacridin-10-ium tetrafluoroborate 97%

Supplier: AMBEED, INC
Warning

A330868-100MG A330868-1G A330868-250MG A330868-50MG
77194-994EA 103 USD InStock
77194-994 77194-996 77194-998 77195-000
9-Mesityl-2,7-dimethyl-10-phenylacridin-10-ium tetrafluoroborate 97%
9-Mesityl-2,7-dimethyl-10-phenylacridin-10-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₃₀H₂₈BF₄N
MDL Number: MFCD28411689
CAS Number: 1621020-00-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty