Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dibenzoylbenzene 98%

Supplier: AMBEED, INC
Warning

A334814-100G A334814-1G A334814-25G A334814-5G
77195-052EA 297 USD InStock
77195-052 77195-054 77195-056 77195-058
1,4-Dibenzoylbenzene 98%
1,4-Dibenzoylbenzene
Precaución: for Research Use Only


Formula: C₂₀H₁₄O₂
MW: 286.33
MDL Number: MFCD00014087
CAS Number: 3016-97-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty