Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,4-Dichlorophenyl)acetonitrile 99%

Supplier: AMBEED, INC
Warning

A336923-100G A336923-10G A336923-25G A336923-5G
77195-096EA 27 USD InStock
77195-096 77195-098 77195-100 77195-102
(3,4-Dichlorophenyl)acetonitrile 99%
(3,4-Dichlorophenyl)acetonitrile
Precaución: for Research Use Only


Formula: C₈H₅Cl₂N
MDL Number: MFCD00001909
CAS Number: 3218-49-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty