Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-((tert-Butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylic acid 96% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A337341-100MG A337341-250MG
77195-108EA 62 USD InStock
77195-108 77195-110
1-((tert-Butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylic acid 96% stabilised
1-((tert-Butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₇NO₄
MDL Number: MFCD14525711
CAS Number: 1001667-24-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty