Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PD-166866 98%

Supplier: AMBEED, INC
Warning

A339638-10MG A339638-1MG A339638-25MG A339638-50MG A339638-5MG
77167-514EA 72 USD InStock
77167-514 77167-516 77167-518 77167-520 77167-522
PD-166866 98%
PD-166866
Precaución: for Research Use Only


Formula: C₂₀H₂₄N₆O₃
MDL Number: MFCD12922514
CAS Number: 192705-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty