Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridine-7-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A341570-100MG A341570-1G A341570-250MG
77167-540EA 13 USD InStock
77167-540 77167-542 77167-544
Imidazo[1,2-a]pyridine-7-carbaldehyde 97%
Imidazo[1,2-a]pyridine-7-carbaldehyde
Precaución: for Research Use Only


Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD09910326
CAS Number: 136117-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty