Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A345260-100MG A345260-10G A345260-1G A345260-250MG A345260-5G
77167-598EA 12 USD InStock
77167-598 77167-600 77167-602 77167-604 77167-606
(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine hydrochloride 98%
(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₈BrClF₃N
MDL Number: MFCD07374367
CAS Number: 336105-43-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty