Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-7-nitro-1H-indole 98%

Supplier: AMBEED, INC
Warning

A346842-100MG A346842-1G A346842-250MG
77195-182EA 20 USD InStock
77195-182 77195-184 77195-186
5-Bromo-7-nitro-1H-indole 98%
5-Bromo-7-nitro-1H-indole
Precaución: For research use only.


Formula: C₈H₅BrN₂O₂
MDL Number: MFCD00619260
CAS Number: 165669-16-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty