Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-1,2,4-triazolide 95%

Supplier: AMBEED, INC
Warning

A354698-1000G A354698-100G A354698-25G A354698-500G A354698-5G
77195-246EA 66 USD InStock
77195-246 77195-248 77195-250 77195-252 77195-254
Sodium-1,2,4-triazolide 95%
Sodium-1,2,4-triazolide
Precaución: for Research Use Only


Formula: C₂H₃N₃
MW: 91.05 g/mol
MDL Number: MFCD00044437
CAS Number: 41253-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty