Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Hydroxyethyl)imidazole 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A356029-10G A356029-1G A356029-25G A356029-5G A356029-500G
77167-758EA 9 USD InStock
77167-758 77167-760 77167-762 77167-764 77684-047
1-(2-Hydroxyethyl)imidazole 97%, Ambeed.Inc
1-(2-Hydroxyethyl)imidazole
Precaución: For research use only.


Formula: C₅H₈N₂O
MDL Number: MFCD00209980
CAS Number: 1615-14-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty