Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-5-nitroaniline 95%

Supplier: AMBEED, INC
Warning

A357139-100MG A357139-250MG A357139-5G A357139-1G
77195-266EA 58 USD InStock
77195-266 77195-268 77746-705 77746-704
3-Methoxy-5-nitroaniline 95%
3-Methoxy-5-nitroaniline
Precaución: for Research Use Only


Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD01103799
CAS Number: 586-10-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty