Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-2,4-xylenol 98%

Supplier: AMBEED, INC
Danger

A362852-1G A362852-25G A362852-5G
77195-366EA 17 USD InStock
77195-366 77195-368 77195-370
6-Amino-2,4-xylenol 98%
6-Amino-2,4-xylenol
Precaución: for Research Use Only


Formula: C₈H₁₁NO
MDL Number: MFCD00007691
CAS Number: 41458-65-5
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty