Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethylphenylboronic acid 98+%

Supplier: AMBEED, INC
Warning

A367002-100G A367002-1G A367002-25G A367002-5G
77195-414EA 204 USD InStock
77195-414 77195-416 77195-418 77195-420
2,3-Dimethylphenylboronic acid 98+%
2,3-Dimethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₁BO₂
MDL Number: MFCD01863524
CAS Number: 183158-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty