Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,2-Dioxathiane-2,2-dioxide 98%

Supplier: AMBEED, INC
Danger

A367205-1G A367205-25G A367205-5G
77195-426EA 7 USD InStock
77195-426 77195-428 77195-430
1,3,2-Dioxathiane-2,2-dioxide 98%
1,3,2-Dioxathiane-2,2-dioxide
Precaución: for Research Use Only


Formula: C₃H₆O₄S
MW: 138.14 g/mol
MDL Number: MFCD00801144
CAS Number: 1073-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty