Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Acetylphenyl)-1H-pyrazole-3-carboxamide 97%

Supplier: AMBEED, INC
Warning

A367574-100MG A367574-1G A367574-250MG
77195-432EA 22 USD InStock
77195-432 77195-434 77195-436
N-(4-Acetylphenyl)-1H-pyrazole-3-carboxamide 97%
N-(4-Acetylphenyl)-1H-pyrazole-3-carboxamide
Precaución: for Research Use Only


Formula: C₁₂H₁₁N₃O₂
MDL Number: MFCD02128528
CAS Number: 305346-13-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty