Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(p-Tolylethynyl)aniline 97%

Supplier: AMBEED, INC
Warning

A369874-10G A369874-1G A369874-250MG A369874-5G
77195-480EA 25.75 USD InStock
77195-480 77195-482 77195-484 77195-486 77195-488
2-(p-Tolylethynyl)aniline 97%
2-(p-Tolylethynyl)aniline
Precaución: for Research Use Only


Formula: C₁₅H₁₃N
MDL Number: MFCD00168854
CAS Number: 124643-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty