Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A372746-100MG A372746-1G A372746-250MG A372746-5G
77167-974EA 15 USD InStock
77167-974 77167-976 77167-978 77167-980
(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98%, ee 99%
(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline)
Precaución: for Research Use Only


Formula: C₁₅H₂₆N₂O₂
MW: 266.39 g/mol
MDL Number: MFCD08458310
CAS Number: 131833-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty