Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(4,5-dimethoxy-2-nitrophenyl)acetate 95%

Supplier: AMBEED, INC
Warning

A374944-100MG A374944-1G A374944-250MG
77168-004EA 24 USD InStock
77168-004 77168-006 77168-008
Methyl 2-(4,5-dimethoxy-2-nitrophenyl)acetate 95%
Methyl 2-(4,5-dimethoxy-2-nitrophenyl)acetate
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₆
MDL Number: MFCD00015617
CAS Number: 2982-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty