Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,4-Dichlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride 97%

Supplier: AMBEED, INC
Warning

A377975-100MG A377975-1G A377975-250MG
77195-572EA 15 USD InStock
77195-572 77195-574 77195-576
1-(3,4-Dichlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride 97%
1-(3,4-Dichlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₄Cl₃NO
MDL Number: MFCD01220371
CAS Number: 75144-12-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty