Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-N,N-dimethylpropan-1-aminium 4-acetamidobenzoate 97%

Supplier: AMBEED, INC
Warning

A380781-100G A380781-10G A380781-25G A380781-5G
77168-054EA 62 USD InStock
77168-054 77168-056 77168-058 77168-060
2-Hydroxy-N,N-dimethylpropan-1-aminium 4-acetamidobenzoate 97%
2-Hydroxy-N,N-dimethylpropan-1-aminium 4-acetamidobenzoate
Precaución: for Research Use Only


Formula: C₁₄H₂₂N₂O₄
MW: 282.33 g/mol
MDL Number: MFCD00867576
CAS Number: 61990-51-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty