Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-4-methylbenzonitrile 97%

Supplier: AMBEED, INC
Warning

A383887-100MG A383887-1G A383887-250MG A383887-5G
77195-702EA 51 USD InStock
77195-702 77195-704 77195-706 77195-708
2,6-Difluoro-4-methylbenzonitrile 97%
2,6-Difluoro-4-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₅F₂N
MDL Number: MFCD28790316
CAS Number: 1803792-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty