Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4,4,4-Trichlorobutanoate 97%

Supplier: AMBEED, INC
Warning

A388362-1G A388362-250MG A388362-5G
77195-780EA 119 USD InStock
77195-780 77195-782 77195-784
Methyl 4,4,4-Trichlorobutanoate 97%
Methyl 4,4,4-Trichlorobutanoate
Precaución: for Research Use Only


Formula: C₅H₇Cl₃O₂
MDL Number: MFCD28144036
CAS Number: 19376-57-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty