Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl spiro[3.3]heptane-2,6-dicarboxylate 95%

Supplier: AMBEED, INC
Warning

A389169-100MG A389169-1G A389169-250MG
77168-178EA 23 USD InStock
77168-178 77168-180 77168-182
Dimethyl spiro[3.3]heptane-2,6-dicarboxylate 95%
Dimethyl spiro[3.3]heptane-2,6-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₄
MDL Number: MFCD11099889
CAS Number: 37942-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty