Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-N3-methylpyridine-2,3-diamine 95%

Supplier: AMBEED, INC
Warning

A389328-100MG A389328-1G A389328-250MG
77168-184EA 37 USD InStock
77168-184 77168-186 77168-188
5-Bromo-N3-methylpyridine-2,3-diamine 95%
5-Bromo-N3-methylpyridine-2,3-diamine
Precaución: for Research Use Only


Formula: C₆H₈BrN₃
MDL Number: MFCD24562466
CAS Number: 166047-15-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty