Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl (3-chlorobenzoyl)acetate 95%

Supplier: AMBEED, INC
Warning

A389759-10G A389759-1G A389759-250MG A389759-5G
77168-192EA 82 USD InStock
77168-192 77168-194 77168-196 77168-198
Ethyl (3-chlorobenzoyl)acetate 95%
Ethyl (3-chlorobenzoyl)acetate
Precaución: for Research Use Only


Formula: C₁₁H₁₁ClO₃
MW: 226.65 g/mol
MDL Number: MFCD03424810
CAS Number: 33167-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty