Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorophthalic anhydride 95%

Supplier: AMBEED, INC
Warning

A390615-100G A390615-25G A390615-500G A390615-5G
77195-792EA 23 USD InStock
77195-792 77195-794 77195-796 77195-798
3-Chlorophthalic anhydride 95%
3-Chlorophthalic anhydride
Precaución: for Research Use Only


Formula: C₈H₃ClO₃
MW: 182.56 g/mol
MDL Number: MFCD00023107
CAS Number: 117-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty