Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-iodonicotinic acid 98%

Supplier: AMBEED, INC
Warning

A393502-1G A393502-250MG A393502-5G
77168-244EA 23 USD InStock
77168-244 77168-246 77168-248
2-Chloro-4-iodonicotinic acid 98%
2-Chloro-4-iodonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
MDL Number: MFCD05663520
CAS Number: 544671-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty