Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrafluoroisophthalic acid 98%

Supplier: AMBEED, INC
Warning

A398677-100MG A398677-1G A398677-250MG A398677-5G
77195-906EA 21 USD InStock
77195-906 77195-908 77195-910 77195-912
Tetrafluoroisophthalic acid 98%
Tetrafluoroisophthalic acid
Precaución: for Research Use Only


Formula: C₈H₂F₄O₄
MW: 238.0927 g/mol
MDL Number: MFCD00042379
CAS Number: 1551-39-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty