Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(bromomethyl)thiazole-5-carboxylate 95%

Supplier: AMBEED, INC
Danger

A398820-100MG A398820-1G A398820-250MG
77168-284EA 144 USD InStock
77168-284 77168-286 77168-288
Ethyl 2-(bromomethyl)thiazole-5-carboxylate 95%
Ethyl 2-(bromomethyl)thiazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₇H₈BrNO₂S
MDL Number: MFCD12913872
CAS Number: 173089-24-2
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty