Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Dimethyl-1H,1'H-2,2'-biimidazole 97%

Supplier: AMBEED, INC
Warning

A399589-100MG A399589-250MG A399589-50MG
77195-916EA 46 USD InStock
77195-916 77195-918 77195-920
1,1'-Dimethyl-1H,1'H-2,2'-biimidazole 97%
1,1'-Dimethyl-1H,1'H-2,2'-biimidazole
Precaución: for Research Use Only


Formula: C₈H₁₀N₄
MW: 162.19 g/mol
MDL Number: MFCD00482464
CAS Number: 37570-94-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty