Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenylbiguanide 98%

Supplier: AMBEED, INC
Warning

A401395-100MG A401395-10G A401395-1G A401395-1MG A401395-25G A401395-5G
77195-932EA 9 USD InStock
77195-932 77195-934 77195-936 77195-938 77195-940 77195-942
1-Phenylbiguanide 98%
1-Phenylbiguanide
Precaución: for Research Use Only


Formula: C₈H₁₁N₅
MW: 177.2064 g/mol
MDL Number: MFCD00179077
CAS Number: 102-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty