Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,4-Dichlorophenyl)cyclopropanecarbonitrile 98%

Supplier: AMBEED, INC
Warning

A402008-1G A402008-250MG A402008-5G
77168-306EA 14 USD InStock
77168-306 77168-308 77168-310
1-(3,4-Dichlorophenyl)cyclopropanecarbonitrile 98%
1-(3,4-Dichlorophenyl)cyclopropanecarbonitrile
Precaución: for Research Use Only


Formula: C₁₀H₇Cl₂N
MDL Number: MFCD09743443
CAS Number: 124276-57-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty