Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Ethyl-2,3-dibromopropionate 98%

Supplier: AMBEED, INC
Danger

A405421-1000G A405421-100G A405421-25G A405421-500G
77168-330EA 241 USD InStock
77168-330 77168-332 77168-334 77168-336
(±)-Ethyl-2,3-dibromopropionate 98%
(±)-Ethyl-2,3-dibromopropionate
Precaución: for Research Use Only


Formula: C₅H₈Br₂O₂
MW: 259.93 g/mol
MDL Number: MFCD00000212
CAS Number: 3674-13-3
UN: 2922
ADR: 8,6.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty