Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium thiophene-2-sulfinate 97%

Supplier: AMBEED, INC
Warning

A405509-100MG A405509-1G A405509-250MG
77196-000EA 59 USD InStock
77196-000 77196-002 77196-004
Sodium thiophene-2-sulfinate 97%
Sodium thiophene-2-sulfinate
Precaución: for Research Use Only


Formula: C₄H₃NaO₂S₂
MDL Number: MFCD11656003
CAS Number: 38945-01-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty