Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (4-chloro-2-methylphenyl)trifluoroborate 98%

Supplier: AMBEED, INC
Warning

A405579-1G A405579-250MG A405579-5G
77196-012EA 150 USD InStock
77196-012 77196-014 77196-016
Potassium (4-chloro-2-methylphenyl)trifluoroborate 98%
Potassium (4-chloro-2-methylphenyl)trifluoroborate
Precaución: for Research Use Only


Formula: C₇H₆BClF₃K
MW: 232.48 g/mol
MDL Number: MFCD09993378
CAS Number: 1620108-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty